Learn More
2,3-Dichlorophenol-OD, CDN
Product Code. 30320519
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.5g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30320519
Brand: CDN CDND79190.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,3-Dichlorophenol-OD
CAS 59457-02-2
Molecular Formula C6 D H3 Cl2 O
Quantity 0.25 g
Synonym Phenol-d, 2,3-dichloro- (9 CI), 2,3-Dichlorophenol-OD, 2,3-Dichlorophenol-d
InChI Formula InChI=1 S/C6H4Cl2O/c7-4-2-1-3-5(9)6(4)8/h1-3,9 H/i/hD
SMILES [2 H]Oc1cccc(Cl)c1Cl
IUPAC Name 1,2-dichloro-3-deuteriooxybenzene
Molecular Weight (g/mol) 164.0075
Formula Weight 162.9702 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity