Learn More
1-Iodopentane-5,5,5-d3, CDN
Product Code. 30318006
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30318006
Brand: CDN CDND64900.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-Iodopentane-5,5,5-D3
CAS 920502-31-4
Molecular Formula C5 D3 H8 I
Quantity 0.25 g
Synonym Pentane-1,1,1-d3, 5-iodo- (ACI), 5-Iodopentane-1,1,1-d3 (ACI), 1-Iodopentane-5,5,5-d3, 1-Iodopentane-5,5,5-D3
InChI Formula InChI=1 S/C5H11I/c1-2-3-4-5-6/h2-5H2,1H3/i1D3
SMILES [2 H]C([2 H])([2 H])CCCCI
IUPAC Name 1,1,1-trideuterio-5-iodopentane
Molecular Weight (g/mol) 201.0638
Formula Weight 201.0094 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity