missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromooctane-1,1-d2, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 1-Bromooctane-1,1-D2 |
| CAS | 86423-34-9 |
| Molecular Formula | C8 D2 H15 Br |
| Quantity | 0.25 g |
| Synonym | Octane-1,1-d2, 1-bromo- (6 CI, 9 CI, ACI), 1-Bromooctane-1,1-d2 (ACI), 1-Bromooctane-1,1-D2, 1-Bromooctane-d2, Octyl bromide-1,1-d2 |
| InChI Formula | InChI=1 S/C8H17Br/c1-2-3-4-5-6-7-8-9/h2-8H2,1H3/i8D2 |
| SMILES | [2 H]C([2 H])(Br)CCCCCCC |
| IUPAC Name | 1-bromo-1,1-dideuteriooctane |
| Molecular Weight (g/mol) | 195.1369 |
| Formula Weight | 194.0639 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction