missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromononane-8,8,9,9,9-d5, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 1-Bromononane-8,8,9,9,9-D5 |
| Molecular Formula | C9 D5 H14 Br |
| Quantity | 0.25 g |
| Synonym | 1-Bromononane-8,8,9,9,9-d5, 9-Bromononane-1,1,1,2,2-d5 |
| InChI Formula | InChI=1 S/C9H19Br/c1-2-3-4-5-6-7-8-9-10/h2-9H2,1H3/i1D3,2D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])CCCCCCCBr |
| IUPAC Name | 9-bromo-1,1,1,2,2-pentadeuteriononane |
| Molecular Weight (g/mol) | 212.182 |
| Formula Weight | 211.0984 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction