Learn More
1-Bromoheptane-d15, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.50g
Specifications
Specifications
| Chemical Name or Material | 1-Bromoheptane-D15 |
| CAS | 98195-42-7 |
| Molecular Formula | C7 D15 Br |
| Quantity | 0.25 g |
| Synonym | Heptane-1,1,1,2,2,3,3,4,4,5,5,6,6,7,7-d15, 7-bromo- (9 CI), Heptyl-d15 bromide, 7-Bromoheptane-1,1,1,2,2,3,3,4,4,5,5,6,6,7,7-d15, 1-Bromoheptane-d15, 1-Bromoheptane-D15 |
| InChI Formula | InChI=1 S/C7H15Br/c1-2-3-4-5-6-7-8/h2-7H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])Br |
| IUPAC Name | 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecadeuterioheptane |
| Molecular Weight (g/mol) | 194.1904 |
| Formula Weight | 193.1299 g/mol |
| Show More |