missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-(1-Bromoethyl-2,2,2-d3)benzene, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | (1-Bromoethyl)benzene-D3 (ethyl-2,2,2-D3) |
| CAS | 23088-42-8 |
| Molecular Formula | C8 D3 H6 Br |
| Quantity | 0.25 g |
| Synonym | Benzene, (1-bromoethyl-2,2,2-d3)- (8 CI, 9 CI, ACI), (1-Bromoethyl-2,2,2-d3)benzene (ACI), (1-Bromoethyl)benzene-D3 (ethyl-2,2,2-D3) |
| InChI Formula | InChI=1 S/C8H9Br/c1-7(9)8-5-3-2-4-6-8/h2-7 H,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])C(Br)c1ccccc1 |
| IUPAC Name | (1-bromo-2,2,2-trideuterioethyl)benzene |
| Molecular Weight (g/mol) | 188.0796 |
| Formula Weight | 187.0076 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction