Learn More
1,9-Nonanedioic-d14 Acid, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Nonanedioic-D14 Acid |
| CAS | 119176-67-9 |
| Molecular Formula | C9 2H14 H2 O4 |
| Quantity | 0.25 g |
| InChI Formula | InChI=1 S/C9H16O4/c10-8(11)6-4-2-1-3-5-7-9(12)13/h1-7H2,(H,10,11)(H,12,13)/i1D2,2D2,3D2,4D2,5D2,6D2,7D2 |
| SMILES | [2 H]C([2 H])(C(=O)O)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)O |
| IUPAC Name | 2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecadeuteriononanedioic acid |
| Molecular Weight (g/mol) | 202.31 |
| Formula Weight | 202.1927 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |