missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3,4-Tetrachlorobenzene-d2, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 1,2,3,4-Tetrachlorobenzene-d2 |
| CAS | 2199-73-7 |
| Molecular Formula | C6 2H2 Cl4 |
| Quantity | 0.25 g |
| Synonym | Benzene-1,2-d2, 3,4,5,6-tetrachloro- (9 CI), Benzene-o-d2, 3,4,5,6-tetrachloro- (8 CI), Benzene-o-d2, tetrachloro- (7 CI), 1,2,3,4-Tetrachlorobenzene D2 |
| InChI Formula | InChI=1 S/C6H2Cl4/c7-3-1-2-4(8)6(10)5(3)9/h1-2 H/i1D,2 D |
| SMILES | [2 H]c1c([2 H])c(Cl)c(Cl)c(Cl)c1Cl |
| IUPAC Name | 1,2,3,4-tetrachloro-5,6-dideuteriobenzene |
| Molecular Weight (g/mol) | 217.9 |
| Formula Weight | 215.9036 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction