Learn More
1,10-Dibromodecane-d20, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 1,10-Dibromodecane-D20 |
| CAS | 150017-88-2 |
| Molecular Formula | C10 D20 Br2 |
| Quantity | 0.25 g |
| Synonym | Decane-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-d20, 1,10-dibromo- (9 CI), 1,10-Dibromodecane-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-d20, 1,10-Dibromodecane-D20, 1,10-Dibromodecane-d20 |
| InChI Formula | InChI=1 S/C10H20Br2/c11-9-7-5-3-1-2-4-6-8-10-12/h1-10H2/i1D2,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2 |
| SMILES | [2 H]C([2 H])(Br)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])Br |
| IUPAC Name | 1,10-dibromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-icosadeuteriodecane |
| Molecular Weight (g/mol) | 320.197 |
| Formula Weight | 318.1187 g/mol |
| Show More |