missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uridine-5,6-d2, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Uridine-5,6-d2 |
| CAS | 40632-21-1 |
| Molecular Formula | C9 2H2 H10 N2 O6 |
| Quantity | 0.1 g |
| InChI Formula | InChI=1 S/C9H12N2O6/c12-3-4-6(14)7(15)8(17-4)11-2-1-5(13)10-9(11)16/h1-2,4,6-8,12,14-15 H,3H2,(H,10,13,16)/t4-,6-,7-,8-/m1/s1/i1D,2 D |
| SMILES | [2 H]C1=C([2 H])C(=O)NC(=O)N1[C@@H]2 O[C@H](CO)[C@@H](O)[C@H]2 O |
| IUPAC Name | 5,6-dideuterio-1-[(2 R,3 R,4 S,5 R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione |
| Molecular Weight (g/mol) | 246.21 |
| Formula Weight | 246.0821 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction