Learn More
Sodium Hexanoate-6,6,6-d3, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
Specifications
Specifications
| Chemical Name or Material | Sodium Hexanoate-6,6,6-D3 |
| CAS | 1219794-91-8 |
| Molecular Formula | C6 D3 H8 O2 . Na |
| Quantity | 0.1 g |
| Synonym | Hexanoic acid-d3 sodium salt, Hexanoic-6,6,6-D3 acid sodium salt, Caproic-ω-d3 acid sodium salt, Sodium hexanoate-6,6,6-D3, Sodium hexanoate-6,6,6-d3, Sodium caproate-ε-d3 |
| InChI Formula | InChI=1 S/C6H12O2.Na/c1-2-3-4-5-6(7)8;/h2-5H2,1H3,(H,7,8);/q;+1/p-1/i1D3; |
| SMILES | [Na+].[2 H]C([2 H])([2 H])CCCCC(=O)[O-] |
| IUPAC Name | sodium;6,6,6-trideuteriohexanoate |
| Molecular Weight (g/mol) | 141.159 |
| Formula Weight | 141.085 g/mol |
| Show More |