Learn More
Sodium DL-Lactate-3,3,3-d3 (60% w/w in H2O), CDN
Product Code. 30320514
Click to view available options
Quantity:
0.1 g
Unit Size:
0.1g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30320514
Brand: CDN CDND65560.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Sodium DL-Lactate-d3
CAS 1219802-24-0
Solvent or Matrix H2O
Molecular Formula C3 2H3 H2 O3 . Na
Quantity 0.1 g
InChI Formula InChI=1 S/C3H6O3.Na/c1-2(4)3(5)6;/h2,4 H,1H3,(H,5,6);/q;+1/p-1/i1D3;
SMILES [Na+].[2 H]C([2 H])([2 H])C(O)C(=O)[O-]
IUPAC Name sodium;3,3,3-trideuterio-2-hydroxypropanoate
Molecular Weight (g/mol) 115.08
Formula Weight 115.0325 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity
Concentration or Composition (by Analyte or Components) 60 %