Learn More
Sodium Butyrate-4,4,4-d3, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Sodium Butanoate-4,4,4-D3 |
| CAS | 347840-05-5 |
| Molecular Formula | C4 D3 H4 O2 . Na |
| Quantity | 0.1 g |
| Synonym | Butanoic-4,4,4-d3 acid, sodium salt (9 CI), Sodium butanoate-4,4,4-D3, Butanoic-4,4,4-d3 acid sodium salt, Sodium butanoate-4,4,4-d3, Sodium butyrate-4,4,4-d3 |
| InChI Formula | InChI=1 S/C4H8O2.Na/c1-2-3-4(5)6;/h2-3H2,1H3,(H,5,6);/q;+1/p-1/i1D3; |
| SMILES | [Na+].[2 H]C([2 H])([2 H])CCC(=O)[O-] |
| IUPAC Name | sodium;4,4,4-trideuteriobutanoate |
| Molecular Weight (g/mol) | 113.105 |
| Formula Weight | 113.053 g/mol |
| Show More |