Learn More
Perylene-d12, CDN
Click to view available options
Quantity:
0.1 g
Unit Size:
0.1g
Specifications
Specifications
| Chemical Name or Material | Perylene D12 |
| CAS | 1520-96-3 |
| Molecular Formula | C20 2H12 |
| Quantity | 0.1 g |
| Synonym | Perylene-1,2,3,4,5,6,7,8,9,10,11,12-d12, Perylene-d12 (7 CI,8 CI,9 CI), Perdeuterioperylene, [2H12]Perylene |
| InChI Formula | InChI=1 S/C20H12/c1-5-13-6-2-11-17-18-12-4-8-14-7-3-10-16(20(14)18)15(9-1)19(13)17/h1-12 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D,10 D,11 D,12 D |
| SMILES | [2 H]c1c([2 H])c2c([2 H])c([2 H])c([2 H])c3c4c([2 H])c([2 H])c([2 H])c5c([2 H])c([2 H])c([2 H])c(c(c1[2 H])c23)c45 |
| IUPAC Name | 1,2,3,4,5,6,7,8,9,10,11,12-dodecadeuterioperylene |
| Molecular Weight (g/mol) | 264.38 |
| Formula Weight | 264.1692 g/mol |
| Show More |