missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinamide-2,4,5,6-d4, CDN
Click to view available options
Quantity:
0.1 g
0.5 g
Unit Size:
0.1g
0.5g
Specifications
Specifications
| Chemical Name or Material | Nicotinamide-2,4,5,6-D4 |
| CAS | 347841-88-7 |
| Molecular Formula | C6 2H4 H2 N2 O |
| Quantity | 0.1 g |
| Synonym | 3-Pyridinecarboxamide-d4, 3-(Aminocarbonyl)pyridine-d4, 3-Amidopyridine-d4, 3-Carbamoylpyridine-d4, 3-Pyridinecarboxylic acid amide-d4, Aminicotin-d4, Benicot-d4, Nicosylamide-d4, Nicotilamide-d4, Nicotinic Acid Amide-d4, Vitamin B-d4, Vitamin B3-d4, Nicotinamide-d4 |
| InChI Formula | InChI=1 S/C6H6N2O/c7-6(9)5-2-1-3-8-4-5/h1-4 H,(H2,7,9)/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1nc([2 H])c(C(=O)N)c([2 H])c1[2 H] |
| IUPAC Name | 2,4,5,6-tetradeuteriopyridine-3-carboxamide |
| Molecular Weight (g/mol) | 126.15 |
| Formula Weight | 126.0731 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction