missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Tetradecyl-d29-amine, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 1-Tetradecanamine-D29 |
| CAS | 204244-82-6 |
| Molecular Formula | C14 D29 H2 N |
| Quantity | 0.1 g |
| Synonym | 1-Tetradecan-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-d29-amine (9 CI), n-Tetradecyl-d29-amine, 1-Tetradecanamine-D29, Tetradecylamine-d29, 1-Aminotetradecane-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-d29, 1-Tetradecylamine-d29, Myristylamine-d29 |
| InChI Formula | InChI=1 S/C14H31N/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15/h2-15H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])N |
| IUPAC Name | 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-nonacosadeuteriotetradecan-1-amine |
| Molecular Weight (g/mol) | 242.581 |
| Formula Weight | 242.428 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction