Learn More
n-Nonyl-6,6,7,7-d4 Alcohol, CDN
Product Code. 30325770
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30325770
Brand: CDN CDND63310.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-Nonanol-6,6,7,7-D4
CAS 33975-46-1
Molecular Formula C9 D4 H16 O
Quantity 0.1 g
Synonym Nonyl-6,6,7,7-d4 alcohol (8 CI), 1-Nonanol-6,6,7,7-D4, Nonanol-6,6,7,7-d4, n-Nonyl-6,6,7,7-d4 alcohol
InChI Formula InChI=1 S/C9H20O/c1-2-3-4-5-6-7-8-9-10/h10H,2-9H2,1H3/i3D2,4D2
SMILES [2 H]C([2 H])(CC)C([2 H])([2 H])CCCCCO
IUPAC Name 6,6,7,7-tetradeuteriononan-1-ol
Molecular Weight (g/mol) 148.279
Formula Weight 148.177 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity