Learn More
N-Nitrosopiperidine-d10, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | N-Nitrosopiperidine-d10 |
| CAS | 960049-21-2 |
| Molecular Formula | C5 2H10 N2 O |
| Quantity | 0.1 g |
| InChI Formula | InChI=1 S/C5H10N2O/c8-6-7-4-2-1-3-5-7/h1-5H2/i1D2,2D2,3D2,4D2,5D2 |
| SMILES | [2 H]C1([2 H])N(N=O)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C1([2 H])[2 H] |
| IUPAC Name | 2,2,3,3,4,4,5,5,6,6-decadeuterio-1-nitrosopiperidine |
| Molecular Weight (g/mol) | 124.21 |
| Formula Weight | 124.1421 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |