missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethyl-d3-chloroacetamide (N-methyl-d3), CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.1g
Specifications
Specifications
| Chemical Name or Material | 2-Chloro-N,N-dimethylacetamide-D3 (N-methyl-D3) |
| CAS | 1219802-19-3 |
| Molecular Formula | C4 D3 H5 Cl N O |
| Quantity | 0.1 g |
| Synonym | 2-Chloro-N-methyl-N-(methyl-d3)acetamide, N-Methyl-N-(methyl-d3)-2-chloroacetamide, α-Chloro-N-methyl-N-(methyl-d3)acetamide, N,N-Dimethyl-d3-chloroacetamide (N-methyl-d3), 2-Chloro-N,N-dimethylacetamide-D3 (N-methyl-D3) |
| InChI Formula | InChI=1 S/C4H8ClNO/c1-6(2)4(7)3-5/h3H2,1-2H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])N(C)C(=O)CCl |
| IUPAC Name | 2-chloro-N-methyl-N-(trideuteriomethyl)acetamide |
| Molecular Weight (g/mol) | 124.584 |
| Formula Weight | 124.048 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction