Learn More
n-Dodecyl-d25-trimethylammonium Bromide, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Dodecyltrimethylammonium-D25 (dodecyl-D25) Bromide |
| CAS | 2309360-05-0 |
| Molecular Formula | C15 D25 H9 N . Br |
| Quantity | 0.1 g |
| Synonym | 1-Dodecan-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-d25-aminium, N,N,N-trimethyl-, bromide (1:1) (ACI), Dodecyltrimethylammonium-d25 bromide, Dodecyl-d25-trimethylammonium bromide, Lauryltrimethylammonium-d25 bromide, Dodecyltrimethylammonium-D25 (dodecyl-D25) bromide, N,N,N-Trimethyl-1-dodecanaminium-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-d25 bromide |
| InChI Formula | InChI=1 S/C15H34N.BrH/c1-5-6-7-8-9-10-11-12-13-14-15-16(2,3)4;/h5-15H2,1-4H3;1 H/q+1;/p-1/i1D3,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2; |
| SMILES | [Br-].[2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[N+](C)(C)C |
| IUPAC Name | trimethyl(1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-pentacosadeuteriododecyl)azanium;bromide |
| Molecular Weight (g/mol) | 333.495 |
| Formula Weight | 332.344 g/mol |
| Show More |