Learn More
n-Decyl-9,9,10,10,10-d5 Alcohol, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 1-Decanol-9,9,10,10,10-D5 |
| CAS | 57368-00-0 |
| Molecular Formula | C10 D5 H17 O |
| Quantity | 0.1 g |
| Synonym | 1-Decan-9,9,10,10,10-d5-ol (9 CI), 9,9,10,10,10-Pentadeuteriodecan-1-ol, 1-Decanol-9,9,10,10,10-d5, n-Decyl-9,9,10,10,10-d5 alcohol |
| InChI Formula | InChI=1 S/C10H22O/c1-2-3-4-5-6-7-8-9-10-11/h11H,2-10H2,1H3/i1D3,2D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])CCCCCCCCO |
| IUPAC Name | 9,9,10,10,10-pentadeuteriodecan-1-ol |
| Molecular Weight (g/mol) | 163.3119 |
| Formula Weight | 163.1984 g/mol |
| Show More |