missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(Carboxymethyl)-N,N,N-trimethyl-d9-ammonium Chloride, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Betaine-D9 (N,N,N-trimethyl-D9) Hydrochloride |
| CAS | 285979-85-3 |
| Molecular Formula | C5 D9 H3 N O2 . Cl |
| Quantity | 0.1 g |
| Synonym | 2-(Tris(trideuteriomethyl)azaniumyl)acetate hydrochloride, N-(Carboxymethyl)-N,N,N-tri(methyl-d3)ammonium chloride, N-(Carboxymethyl)-N,N,N-trimethyl-d9-ammonium Chloride, Betaine-d9 hydrochloride |
| InChI Formula | InChI=1 S/C5H11NO2.ClH/c1-6(2,3)4-5(7)8;/h4H2,1-3H3;1 H/i1D3,2D3,3D3; |
| SMILES | [Cl-].[2 H]C([2 H])([2 H])[N+](CC(=O)O)(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | carboxymethyl-tris(trideuteriomethyl)azanium;chloride |
| Molecular Weight (g/mol) | 162.6627 |
| Formula Weight | 162.1121 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction