Learn More
N-Benzoyl-d5-glycine, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Hippuric Acid-d5 |
| CAS | 53518-98-2 |
| Molecular Formula | C9 2H5 H4 N O3 |
| Quantity | 0.1 g |
| InChI Formula | InChI=1 S/C9H9NO3/c11-8(12)6-10-9(13)7-4-2-1-3-5-7/h1-5 H,6H2,(H,10,13)(H,11,12)/i1D,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)NCC(=O)O)c([2 H])c1[2 H] |
| IUPAC Name | 2-[(2,3,4,5,6-pentadeuteriobenzoyl)amino]acetic acid |
| Molecular Weight (g/mol) | 184.2 |
| Formula Weight | 184.0896 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |