Learn More
N-Acetyl-D-glutamic-2,3,3,4,4-d5 Acid, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | N-Acetyl-D-glutamic-2,3,3,4,4-D5 Acid |
| CAS | 14341-87-8 |
| Molecular Formula | C7 D5 H6 N O5 |
| Quantity | 0.1 g |
| Synonym | Glutamic-2,3,3,4,4-d5 acid, N-acetyl-, D- (8 CI), N-Acetyl-D-glutamic-2,3,3,4,4-d5 acid, (2 R)-2-Acetamidopentanedioic-2,3,3,4,4-d5 acid, N-Acetyl-D-glutamic-2,3,3,4,4-D5 acid |
| InChI Formula | InChI=1 S/C7H11NO5/c1-4(9)8-5(7(12)13)2-3-6(10)11/h5H,2-3H2,1H3,(H,8,9)(H,10,11)(H,12,13)/t5-/m1/s1/i2D2,3D2,5 D |
| SMILES | [2 H]C([2 H])(C(=O)O)C([2 H])([2 H])[C@@]([2 H])(NC(=O)C)C(=O)O |
| IUPAC Name | (2 R)-2-acetamido-2,3,3,4,4-pentadeuteriopentanedioic acid |
| Molecular Weight (g/mol) | 194.197 |
| Formula Weight | 194.095 g/mol |
| Show More |