missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(4-Methyl-d3-benzoyl-d4)glycine, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 4-Methyl Hippuric Acid-d7 |
| CAS | 1216588-60-1 |
| Molecular Formula | C10 D7 H4 N O3 |
| Quantity | 0.1 g |
| Synonym | 2-[[2,3,5,6-Tetradeuterio-4-(trideuteriomethyl)benzoyl]amino]acetic acid, N-(4-Methylbenzoyl)glycine-D7 (methylbenzoyl-D7), N-(4-Methylbenzoyl)glycine-d7, 4-Methylhippuric acid-d7, 2-[[4-(Methyl-d3)benzoyl-2,3,5,6-d4]amino]acetic acid, p-Methylhippuric acid-d7, p-Toluric acid-d7, 4-Methyl hippuric acid-d7 |
| InChI Formula | InChI=1 S/C10H11NO3/c1-7-2-4-8(5-3-7)10(14)11-6-9(12)13/h2-5 H,6H2,1H3,(H,11,14)(H,12,13)/i1D3,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c([2 H])c1C(=O)NCC(=O)O)C([2 H])([2 H])[2 H] |
| IUPAC Name | 2-[[2,3,5,6-tetradeuterio-4-(trideuteriomethyl)benzoyl]amino]acetic acid |
| Molecular Weight (g/mol) | 200.242 |
| Formula Weight | 200.118 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction