Learn More
Methyl trans-Cinnamate-d7, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Methyl trans-Cinnamate-D7 |
| CAS | 2022152-96-9 |
| Molecular Formula | C10 D7 H3 O2 |
| Quantity | 0.1 g |
| Synonym | 2-Propenoic-2,3-d2 acid, 3-(phenyl-2,3,4,5,6-d5)-, methyl ester, (2 E)- (ACI), Methyl trans-Cinnamate-D7, (2 E)-3-(Phenyl-d5)-2-propenoic-2,3-d2 acid methyl ester, (E)-Cinnamic-d7 acid methyl ester |
| InChI Formula | InChI=1 S/C10H10O2/c1-12-10(11)8-7-9-5-3-2-4-6-9/h2-8 H,1H3/b8-7+/i2D,3 D,4 D,5 D,6 D,7 D,8 D |
| SMILES | [2 H]\C(=C(\[2 H])/c1c([2 H])c([2 H])c([2 H])c([2 H])c1[2 H])\C(=O)OC |
| IUPAC Name | methyl (E)-2,3-dideuterio-3-(2,3,4,5,6-pentadeuteriophenyl)prop-2-enoate |
| Molecular Weight (g/mol) | 169.2283 |
| Formula Weight | 169.112 g/mol |
| Show More |