Learn More
Methyl 3-Bromopropionate-2,2,3,3-d4, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Methyl 3-bromopropanoate-2,2,3,3-D4 |
| CAS | 1219803-82-3 |
| Molecular Formula | C4 D4 H3 Br O2 |
| Quantity | 0.1 g |
| Synonym | Methyl 3-bromopropanoate-2,2,3,3-D4, 3-Bromopropanoic-2,2,3,3-d4 acid methyl ester, 3-Bromopropionic-2,2,3,3-d4 acid methyl ester, Methyl 3-bromopropionate-2,2,3,3-d4 |
| InChI Formula | InChI=1 S/C4H7BrO2/c1-7-4(6)2-3-5/h2-3H2,1H3/i2D2,3D2 |
| SMILES | [2 H]C([2 H])(Br)C([2 H])([2 H])C(=O)OC |
| IUPAC Name | methyl 3-bromo-2,2,3,3-tetradeuteriopropanoate |
| Molecular Weight (g/mol) | 171.0258 |
| Formula Weight | 169.988 g/mol |
| Show More |