missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Proline-2,5,5-d3, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | L-Proline-2,5,5-d3 |
| CAS | 65807-22-9 |
| Molecular Formula | C5 D3 H6 N O2 |
| Quantity | 0.1 g |
| InChI Formula | InChI=1 S/C5H9NO2/c7-5(8)4-2-1-3-6-4/h4,6 H,1-3H2,(H,7,8)/t4-/m0/s1/i3D2,4 D |
| SMILES | [2 H]C1([2 H])CC[C@]([2 H])(N1)C(=O)O |
| IUPAC Name | (2 S)-2,5,5-trideuteriopyrrolidine-2-carboxylic acid |
| Molecular Weight (g/mol) | 118.149 |
| Formula Weight | 118.0822 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction