Learn More
L-Phenyl-d5-alanine-2,3,3-d3, CDN
Product Code. 30317968
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30317968
Brand: CDN CDND12410.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material L-Phenyl-d5-alanine-2,3,3-d3
CAS 17942-32-4
Molecular Formula C9 D8 H3 N O2
Quantity 0.1 g
Synonym L-Phenyl-2,3,4,5,6-d5-alanine-α,β,β-d3, L-Phenylalanine-d8
InChI Formula InChI=1 S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8 H,6,10H2,(H,11,12)/t8-/m0/s1/i1D,2 D,3 D,4 D,5 D,6D2,8 D
SMILES [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])C([2 H])([2 H])[C@]([2 H])(N)C(=O)O
IUPAC Name (2 S)-2-amino-2,3,3-trideuterio-3-(2,3,4,5,6-pentadeuteriophenyl)propanoic acid
Molecular Weight (g/mol) 173.24
Formula Weight 173.1292 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity