missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Leucine-d10, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | L-Leucine-2,3,3,4,5,5,5,5',5',5'-d10 |
| CAS | 106972-44-5 |
| Molecular Formula | C6 2H10 H3 N O2 |
| Quantity | 0.1 g |
| Synonym | L-Leucine-2,3,3,4,5,5,5,5',5',5'-d10, L-Leucine-d10, (S)-2-Amino-(2,3,3,4,5,5,5-heptadeuterio)-4-(trideuteriomethyl)pentanoic acid |
| InChI Formula | InChI=1 S/C6H13NO2/c1-4(2)3-5(7)6(8)9/h4-5 H,3,7H2,1-2H3,(H,8,9)/t5-/m0/s1/i1D3,2D3,3D2,4 D,5 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[C@]([2 H])(N)C(=O)O |
| IUPAC Name | (2 S)-2-amino-2,3,3,4,5,5,5-heptadeuterio-4-(trideuteriomethyl)pentanoic acid |
| Molecular Weight (g/mol) | 141.23 |
| Formula Weight | 141.1574 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction