missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexanal-d12, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Hexanal-d12 |
| CAS | 1219803-74-3 |
| Molecular Formula | C6 2H12 O |
| Quantity | 0.1 g |
| InChI Formula | InChI=1 S/C6H12O/c1-2-3-4-5-6-7/h6H,2-5H2,1H3/i1D3,2D2,3D2,4D2,5D2,6 D |
| SMILES | [2 H]C(=O)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,2,2,3,3,4,4,5,5,6,6,6-dodecadeuteriohexan-1-one |
| Molecular Weight (g/mol) | 112.23 |
| Formula Weight | 112.1641 g/mol |
| Percent Purity | 98 atom % D, min 96% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction