missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexadecanoic-11,11-d2 Acid, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Specifications
Specifications
| Chemical Name or Material | Hexadecanoic-11,11-D2 Acid |
| CAS | 1219805-64-7 |
| Molecular Formula | C16 D2 H30 O2 |
| Quantity | 0.1 g |
| Synonym | Hexadecanoic-11,11-d2 acid, Palmitic-11,11-d2 acid, 11,11-Dideuteriohexadecanoic acid, 11,11-D2-Palmitic acid, Hexadecanoic-11,11-D2 Acid, [11,11-2H2]Hexadecanoic acid, [11,11-2H2]Palmitic acid, Hexadecanoic Acid-11,11-d2 |
| InChI Formula | InChI=1 S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)/i6D2 |
| SMILES | [2 H]C([2 H])(CCCCC)CCCCCCCCCC(=O)O |
| IUPAC Name | 11,11-dideuteriohexadecanoic acid |
| Molecular Weight (g/mol) | 258.4364 |
| Formula Weight | 258.2528 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction