missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl-d6 Phthalate, CDN
Click to view available options
Quantity:
0.1 g
0.5 g
Unit Size:
0.1g
0.5g
Specifications
Specifications
| Chemical Name or Material | Phthalic Acid Di(methyl-d3) Ester |
| CAS | 85448-30-2 |
| Molecular Formula | C10 D6 H4 O4 |
| Quantity | 0.1 g |
| Synonym | 1,2-Benzenedicarboxylic Acid Di(methyl-d3) Ester, 1,2-Benzenedicarboxylic acid, di(methyl-d3) ester (9 CI), Phthalic acid, di(methyl-d3) ester (7 CI), 1,2-Di(methyl-d3) 1,2-benzenedicarboxylate, Phthalic acid dimethyl ester D6, Phthalic acid dimethyl ester-d6, Phthalic acid dimethyl ester D6 (dimethyl-D6), Dimethyl-d6 phthalate, Phthalic acid, bis-methyl ester D6 |
| InChI Formula | InChI=1 S/C10H10O4/c1-13-9(11)7-5-3-4-6-8(7)10(12)14-2/h3-6 H,1-2H3/i1D3,2D3 |
| SMILES | [2 H]C([2 H])([2 H])OC(=O)c1ccccc1C(=O)OC([2 H])([2 H])[2 H] |
| IUPAC Name | bis(trideuteriomethyl) benzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 200.22 |
| Formula Weight | 200.0956 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction