missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl-1,1,1',1'-d4-amine, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Diethylamine-1,1,1',1'-D4 |
| CAS | 60455-40-5 |
| Molecular Formula | C4 D4 H7 N |
| Quantity | 0.1 g |
| Synonym | Ethan-1,1-d2-amine, N-(ethyl-1,1-d2)- (9 CI), 1,1-Dideuterio-N-(1,1-dideuterioethyl)ethanamine, Diethyl-d4-amine, Diethyl-1,1,1',1'-d4-amine, N-(Ethyl-1,1-d2)ethan-1,1-d2-amine, Diethylamine-1,1,1',1'-D4, Diethylamine-1,1,1',1'-D4 |
| InChI Formula | InChI=1 S/C4H11N/c1-3-5-4-2/h5H,3-4H2,1-2H3/i3D2,4D2 |
| SMILES | [2 H]C([2 H])(C)NC([2 H])([2 H])C |
| IUPAC Name | 1,1-dideuterio-N-(1,1-dideuterioethyl)ethanamine |
| Molecular Weight (g/mol) | 77.1615 |
| Formula Weight | 77.1143 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction