missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-n-propyl-d14-amine, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Dipropylamine-D14 |
| CAS | 345909-05-9 |
| Molecular Formula | C6 D14 H N |
| Quantity | 0.1 g |
| Synonym | 1-Propan-1,1,2,2,3,3,3-d7-amine, N-(propyl-d7)- (9 CI), N-(Propyl-d7)-1-propan-1,1,2,2,3,3,3-d7-amine, Dipropylamine-d14, Dipropylamine-D14, Dipropylamine-1,1,1',1',2,2,2',2',3,3,3,3',3',3'-D14, Di-n-propyl-d14-amine |
| InChI Formula | InChI=1 S/C6H15N/c1-3-5-7-6-4-2/h7H,3-6H2,1-2H3/i1D3,2D3,3D2,4D2,5D2,6D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])NC([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,1,2,2,3,3,3-heptadeuterio-N-(1,1,2,2,3,3,3-heptadeuteriopropyl)propan-1-amine |
| Molecular Weight (g/mol) | 115.2763 |
| Formula Weight | 115.2083 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction