missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-n-heptyl Phthalate-3,4,5,6-d4, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Phthalic acid, bis-n-heptyl ester D4 (3,4,5,6 D4) |
| CAS | 358731-41-6 |
| Molecular Formula | C22 2H4 H30 O4 |
| Quantity | 0.1 g |
| InChI Formula | InChI=1 S/C22H34O4/c1-3-5-7-9-13-17-25-21(23)19-15-11-12-16-20(19)22(24)26-18-14-10-8-6-4-2/h11-12,15-16 H,3-10,13-14,17-18H2,1-2H3/i11D,12 D,15 D,16 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)OCCCCCCC)c(C(=O)OCCCCCCC)c1[2 H] |
| IUPAC Name | diheptyl 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 366.53 |
| Formula Weight | 366.2708 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction