Learn More
Di-n-dodecyl Phthalate-3,4,5,6-d4, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.10g
Specifications
Specifications
| Chemical Name or Material | Didodecyl Phthalate-3,4,5,6-D4 |
| CAS | 2708280-31-1 |
| Molecular Formula | C32 D4 H50 O4 |
| Quantity | 0.1 g |
| Synonym | Di-n-dodecyl Phthalate-3,4,5,6-d4, Didodecyl phthalate-3,4,5,6-d4, Dilauryl phthalate-d4, Phthalic Acid-d4 Bis-dodecyl Ester, Didodecyl Phthalate-3,4,5,6-D4 |
| InChI Formula | InChI=1 S/C32H54O4/c1-3-5-7-9-11-13-15-17-19-23-27-35-31(33)29-25-21-22-26-30(29)32(34)36-28-24-20-18-16-14-12-10-8-6-4-2/h21-22,25-26 H,3-20,23-24,27-28H2,1-2H3/i21D,22 D,25 D,26 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)OCCCCCCCCCCCC)c(C(=O)OCCCCCCCCCCCC)c1[2 H] |
| IUPAC Name | didodecyl 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 506.7934 |
| Formula Weight | 506.4273 g/mol |
| Show More |