missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di(n-butyl-d9) 1,10-Decanedioate, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Dibutyl Sebacate-D18 (dibutyl-D18) |
| Molecular Formula | C18 D18 H16 O4 |
| Quantity | 0.1 g |
| Synonym | Decanedioic acid di(butyl-d9) ester, Sebacic acid di(butyl-d9) ester, Di(butyl-d9) decanedioate, Di(butyl-d9) sebacate, Sebacic acid di(n-butyl-d9) ester, Dibutyl Sebacate-d18, Di(n-butyl-d9) 1,10-Decanedioate, Dibutyl Sebacate-D18 (dibutyl-D18) |
| InChI Formula | InChI=1 S/C18H34O4/c1-3-5-15-21-17(19)13-11-9-7-8-10-12-14-18(20)22-16-6-4-2/h3-16H2,1-2H3/i1D3,2D3,3D2,4D2,5D2,6D2,15D2,16D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])OC(=O)CCCCCCCCC(=O)OC([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | bis(1,1,2,2,3,3,4,4,4-nonadeuteriobutyl) decanedioate |
| Molecular Weight (g/mol) | 332.5711 |
| Formula Weight | 332.3587 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction