missing translation for 'onlineSavingsMsg'
Learn More

Deoxycholic Acid-24-13C, CDN

Product Code. 30312869
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
This item is not returnable. View return policy

Product Code. 30312869

Brand: CDN CDNC40700.1G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Deoxycholic Acid (24-13 C)
CAS 52886-37-0
Molecular Formula 13 C C23 H40 O4
Quantity 0.1 g
Synonym Cholan-24-oic-24-13 C acid, 3,12-dihydroxy-, (3α,5β,12α)-, Deoxycholic-24-13 C acid, (3α,5β,12α)-3,12-Dihydroxycholan-24-oic-24-13 C acid, Deoxycholic acid-13 C, Deoxycholic-24-13 C acid, Deoxycholic acid (24-13 C), Deoxycholic acid (24-13 C)
InChI Formula InChI=1 S/C24H40O4/c1-14(4-9-22(27)28)18-7-8-19-17-6-5-15-12-16(25)10-11-23(15,2)20(17)13-21(26)24(18,19)3/h14-21,25-26 H,4-13H2,1-3H3,(H,27,28)/t14-,15-,16-,17+,18-,19+,20+,21+,23+,24-/m1/s1/i22+1
SMILES C[C@H](CC[13 C](=O)O)[C@H]1 CC[C@H]2 [C@@H]3 CC[C@@H]4 C[C@H](O)CC[C@]4(C)[C@H]3 C[C@H](O)[C@]12 C
IUPAC Name (4 R)-4-[(3 R,5 R,8 R,9 S,10 S,12 S,13 R,14 S,17 R)-3,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1 H-cyclopenta[a]phenanthren-17-yl](1 ^{13}C)pentanoic acid
Molecular Weight (g/mol) 393.565
Formula Weight 393.296 g/mol
Percent Purity 99 atom % 13 C, min 98% Chemical Purity
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.