missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Mannitol-1,1,2,3,4,5,6,6-d8, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | D-Mannitol-d8 |
| Molecular Formula | C6 D8 H6 O6 |
| Quantity | 0.1 g |
| Synonym | D-Mannitol-1,1,2,3,4,5,6,6-C-d8, D-Mannitol-1,1,2,3,4,5,6,6-d8, D-Mannitol-d8, D-Mannitol-D8 |
| InChI Formula | InChI=1 S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12 H,1-2H2/t3-,4-,5-,6-/m1/s1/i1D2,2D2,3 D,4 D,5 D,6 D |
| SMILES | [2 H]C([2 H])(O)[C@@]([2 H])(O)[C@@]([2 H])(O)[C@]([2 H])(O)[C@]([2 H])(O)C([2 H])([2 H])O |
| IUPAC Name | (2 R,3 R,4 R,5 R)-1,1,2,3,4,5,6,6-octadeuteriohexane-1,2,3,4,5,6-hexol |
| Molecular Weight (g/mol) | 190.2211 |
| Formula Weight | 190.1293 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction