Learn More
Cyclopropylmethyl-d2 Alcohol, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Cyclopropylmethanol-1,1-D2 |
| CAS | 90568-07-3 |
| Molecular Formula | C4 D2 H6 O |
| Quantity | 0.1 g |
| Synonym | Cyclopropanemethan-d2-ol (6 CI, 9 CI, ACI), Cyclopropanemethan-α,α-d2-ol (7 CI), Cyclopropylmethanol-1,1-D2, Cyclopropylmethyl-d2 Alcohol |
| InChI Formula | InChI=1 S/C4H8O/c5-3-4-1-2-4/h4-5 H,1-3H2/i3D2 |
| SMILES | [2 H]C([2 H])(O)C1CC1 |
| IUPAC Name | cyclopropyl(dideuterio)methanol |
| Molecular Weight (g/mol) | 74.118 |
| Formula Weight | 74.0701 g/mol |
| Show More |