missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric-2,2,4,4-d4 Acid, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Citric Acid-2,2,4,4-d4 |
| CAS | 147664-83-3 |
| Molecular Formula | C6 2H4 H4 O7 |
| Quantity | 0.1 g |
| InChI Formula | InChI=1 S/C6H8O7/c7-3(8)1-6(13,5(11)12)2-4(9)10/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)/i1D2,2D2 |
| SMILES | [2 H]C([2 H])(C(=O)O)C(O)(C(=O)O)C([2 H])([2 H])C(=O)O |
| IUPAC Name | 1,1,3,3-tetradeuterio-2-hydroxypropane-1,2,3-tricarboxylic acid |
| Molecular Weight (g/mol) | 196.15 |
| Formula Weight | 196.0521 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction