missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(3,7-dimethyl-1-octyl) Phthalate-3,4,5,6-d4, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Phthalic Acid Bis(3,7-dimethyloctyl) Ester-d4 |
| CAS | 1398065-81-0 |
| Molecular Formula | C28 D4 H42 O4 |
| Quantity | 0.1 g |
| Synonym | 1,2-Benzenedicarboxylic-3,4,5,6-d4 acid bis(3,7-dimethyloctyl) ester, 1,2-Bis(3,7-dimethyloctyl) 1,2-benzenedicarboxylate-3,4,5,6-d4, Phthalic-d4 acid bis(3,7-dimethyloctyl) ester, Bis(3,7-dimethyloctyl) phthalate-3,4,5,6-d4, Phthalic Acid Bis(3,7-dimethyloctyl) Ester-D4 (phthalic-D4) |
| InChI Formula | InChI=1 S/C28H46O4/c1-21(2)11-9-13-23(5)17-19-31-27(29)25-15-7-8-16-26(25)28(30)32-20-18-24(6)14-10-12-22(3)4/h7-8,15-16,21-24 H,9-14,17-20H2,1-6H3/i7D,8 D,15 D,16 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)OCCC(C)CCCC(C)C)c(C(=O)OCCC(C)CCCC(C)C)c1[2 H] |
| IUPAC Name | bis(3,7-dimethyloctyl) 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 450.6871 |
| Formula Weight | 450.3647 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction