missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl 2-Hydroxybenzoate-3,4,5,6-d4, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Benzyl Salicylate-d4 |
| CAS | 1219802-40-0 |
| Molecular Formula | C14 D4 H8 O3 |
| Quantity | 0.1 g |
| Synonym | 6-Hydroxybenzoic-2,3,4,5-d4 acid phenylmethyl ester, 2-Hydroxybenzoic-3,4,5,6-d4 acid benzyl ester, Salicylic-d4 acid benzyl ester, Salicylic Acid Benzyl Ester-d4, Salicylic Acid Benzyl Ester-D4 (salicyl-D4), Benzyl Salicylate-d4, Benzyl 2-Hydroxybenzoate-3,4,5,6-d4 |
| InChI Formula | InChI=1 S/C14H12O3/c15-13-9-5-4-8-12(13)14(16)17-10-11-6-2-1-3-7-11/h1-9,15 H,10H2/i4D,5 D,8 D,9 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)OCc2ccccc2)c(O)c1[2 H] |
| IUPAC Name | benzyl 2,3,4,5-tetradeuterio-6-hydroxybenzoate |
| Molecular Weight (g/mol) | 232.2679 |
| Formula Weight | 232.1038 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction