missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl-2,3,4,5,6-d5-dimethyl-n-tetradecylammonium Chloride, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Benzyldimethyltetradecylammonium D5 (phenyl D5) Chloride |
| Molecular Formula | C23 D5 H37 N . Cl |
| Quantity | 0.1 g |
| Synonym | Benzyldimethyltetradecylammonium D5 (phenyl D5) Chloride, N-Tetradecyl-N,N-dimethylbenzene-d5-methanaminium chloride, (Benzyl-2,3,4,5-d5)dimethyltetradecylammonium chloride |
| InChI Formula | InChI=1 S/C23H42N.ClH/c1-4-5-6-7-8-9-10-11-12-13-14-18-21-24(2,3)22-23-19-16-15-17-20-23;/h15-17,19-20 H,4-14,18,21-22H2,1-3H3;1 H/q+1;/p-1/i15D,16 D,17 D,19 D,20 D; |
| SMILES | [Cl-].[2 H]c1c([2 H])c([2 H])c(C[N+](C)(C)CCCCCCCCCCCCCC)c([2 H])c1[2 H] |
| IUPAC Name | dimethyl-[(2,3,4,5,6-pentadeuteriophenyl)methyl]-tetradecylazanium;chloride |
| Molecular Weight (g/mol) | 373.0701 |
| Formula Weight | 372.332 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction