Learn More
Azobenzene-d10, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 1,2-Diphenyldiazene-D10 |
| CAS | 30504-49-5 |
| Molecular Formula | C12 D10 N2 |
| Quantity | 0.1 g |
| Synonym | Diazene, 1,2-di(phenyl-d5)- (ACI), 1,2-Di(phenyl-d5)diazene (ACI), Azobenzene-d10 (8 CI), Diazene, di(phenyl-d5)- (9 CI), 1,2-Diphenyldiazene-D10 |
| InChI Formula | InChI=1 S/C12H10N2/c1-3-7-11(8-4-1)13-14-12-9-5-2-6-10-12/h1-10 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D,10 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(N=Nc2c([2 H])c([2 H])c([2 H])c([2 H])c2[2 H])c([2 H])c1[2 H] |
| IUPAC Name | bis(2,3,4,5,6-pentadeuteriophenyl)diazene |
| Molecular Weight (g/mol) | 192.2828 |
| Formula Weight | 192.1472 g/mol |
| Show More |