missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetylcholine-1,1,2,2-d4 Bromide, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Acetylcholine-1,1,2,2-D4 Bromide |
| CAS | 93449-31-1 |
| Molecular Formula | C7 D4 H12 N O2 . Br |
| Quantity | 0.1 g |
| Synonym | Ethan-1,1,2,2-d4-aminium, 2-(acetyloxy)-N,N,N-trimethyl-, bromide (9 CI), 2-(Acetyloxy)-N,N,N-trimethylethan-1,1,2,2-d4-aminium bromide (1:1), Acetylcholine-1,1,2,2-d4 Bromide, Acetylcholine-d4 Bromide, Acetylcholine-1,1,2,2-D4 Bromide |
| InChI Formula | InChI=1 S/C7H16NO2.BrH/c1-7(9)10-6-5-8(2,3)4;/h5-6H2,1-4H3;1 H/q+1;/p-1/i5D2,6D2; |
| SMILES | [Br-].[2 H]C([2 H])(OC(=O)C)C([2 H])([2 H])[N+](C)(C)C |
| IUPAC Name | (2-acetyloxy-1,1,2,2-tetradeuterioethyl)-trimethylazanium;bromide |
| Molecular Weight (g/mol) | 230.1361 |
| Formula Weight | 229.0615 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction