missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Anthracene-d9-carboxylic Acid, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.1g
Specifications
Specifications
| Chemical Name or Material | 9-Anthracenecarboxylic Acid-D9 |
| CAS | 1219803-78-7 |
| Molecular Formula | C15 D9 H O2 |
| Quantity | 0.1 g |
| Synonym | 9-Anthracene-d9-carboxylic Acid, 10-Anthracenecarboxylic-1,2,3,4,5,6,7,8,9-d9 acid, 9-Anthracenecarboxylic-d9 acid, 9-Anthracenecarboxylic acid-D9, 9-Anthroic acid-d9, 9-Carboxyanthracene-d9 |
| InChI Formula | InChI=1 S/C15H10O2/c16-15(17)14-12-7-3-1-5-10(12)9-11-6-2-4-8-13(11)14/h1-9 H,(H,16,17)/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c2c(C(=O)O)c3c([2 H])c([2 H])c([2 H])c([2 H])c3c([2 H])c2c1[2 H] |
| IUPAC Name | 1,2,3,4,5,6,7,8,10-nonadeuterioanthracene-9-carboxylic acid |
| Molecular Weight (g/mol) | 231.2942 |
| Formula Weight | 231.1246 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction