missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Piperidone-3,3,5,5-d4-N-t-BOC, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 1-(tert-Butoxycarbonyl)-4-piperidone-3,3,5,5-D4 |
| CAS | 1246342-66-4 |
| Molecular Formula | C10 D4 H13 N O3 |
| Quantity | 0.1 g |
| Synonym | 1-Piperidine-3,3,5,5-d4-carboxylic acid, 4-oxo-, 1,1-dimethylethyl ester (ACI), 1,1-Dimethylethyl 4-oxo-1-piperidine-3,3,5,5-d4-carboxylate (ACI), 1-(tert-Butoxycarbonyl)-4-piperidinone-3,3,5,5-d4, 1-(tert-Butoxycarbonyl)-4-piperidone-3,3,5,5-D4, N-(tert-Butyloxycarbonyl)-4-piperidone-3,3,5,5-d4, N-Boc-piperidin-4-one-3,3,5,5-d4, 1-t-Boc-4-piperidone-3,3,5,5-d4, 4-Piperidone-3,3,5,5-d4-N-t-BOC |
| InChI Formula | InChI=1 S/C10H17NO3/c1-10(2,3)14-9(13)11-6-4-8(12)5-7-11/h4-7H2,1-3H3/i4D2,5D2 |
| SMILES | [2 H]C1([2 H])CN(CC([2 H])([2 H])C1=O)C(=O)OC(C)(C)C |
| IUPAC Name | tert-butyl 3,3,5,5-tetradeuterio-4-oxopiperidine-1-carboxylate |
| Molecular Weight (g/mol) | 203.2715 |
| Formula Weight | 203.146 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction