missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Oxo-2,2,6,6-tetramethylpiperidine-d16;1-15N-1-oxyl, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 2,2,6,6-Tetramethyl-4-oxopiperidine-d16,1-15 N-1-oxyl |
| CAS | 80404-14-4 |
| Molecular Formula | C9 2H16 15 N O2 |
| Quantity | 0.1 g |
| Synonym | 1-Piperidinyl-3,3,5,5-d4-1-15 N-oxy, 2,2,6,6-tetra(methyl-d3)-4-oxo-, 2,2,6,6-Tetra(methyl-d3)-4-oxo-1-piperidinyl-3,3,5,5-d4-1-15 N-oxy, 4-Oxo-TEMPO-d16,15 N, 4-Oxo-2,2,6,6-tetramethylpiperidine-d16,1-15 N-1-oxyl, 2,2,6,6-Tetramethyl-4-oxopiperidine-d16,1-15 N-1-oxyl, 3,3,5,5-Tetradeuterio-1-λ5-oxidanyl-2,2,6,6-tetrakis(trideuteriomethyl)piperidin-4-one-15 N, 3,3,5,5-Tetradeuterio-1-λ5-oxidanyl-2,2,6,6-tetrakis(trideuteriomethyl)piperidin-4-one-15 N, 3,3,5,5-Tetradeuterio-1-λ5-oxidanyl-2,2,6,6-tetrakis(trideuteriomethyl)piperidin-4-one-15 N |
| InChI Formula | InChI=1 S/C9H16NO2/c1-8(2)5-7(11)6-9(3,4)10(8)12/h5-6H2,1-4H3/i1D3,2D3,3D3,4D3,5D2,6D2,10+1 |
| SMILES | [2 H]C([2 H])([2 H])C1([15 N]([O])C(C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])C(=O)C1([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 3,3,5,5-tetradeuterio-1-λ^{1}-oxidanyl-2,2,6,6-tetrakis(trideuteriomethyl)(1 ^{15}N)azinan-4-one |
| Molecular Weight (g/mol) | 187.32 |
| Formula Weight | 187.2156 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction